Thermo Scientific Chemicals

Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206512500
250 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium phosphate, dibasic heptahydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206512500
250 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206512500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7782-85-6
IUPAC Name
disodium phosphoric acid heptahydrate
Molecular Formula
H17Na2O11P
InChI Key
PYLIXCKOHOHGKQ-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].OP(O)(O)=O
Iron (Fe)
=<5 ppm
Heavy metals (as Pb)
=<5 ppm
Arsenic (As)
=<5 ppm
Appearance (Form)
Crystals
Sulfate (SO4)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text