Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 206520025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium phosphate, tribasic dodecahydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 206520025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206520025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10101-89-0
IUPAC Name
trisodium dodecahydrate phosphate
Molecular Formula
H24Na3O16P
InChI Key
ASTWEMOBIXQPPV-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=O
Chloride (Cl)
=<100 ppm
Appearance (Form)
Powder or crystals or needles
Free alkali (NaOH)
=<2.5 %
Sulfate (SO4)
=<100 ppm
Appearance (Color)
Colorless to white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text