Pierce™ Dimethylformamide (DMF), Sequencing grade
Pierce™ Dimethylformamide (DMF), Sequencing grade
Thermo Scientific™

Pierce™ Dimethylformamide (DMF), Sequencing grade

Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLCRead more
Have Questions?
Catalog NumberQuantity
2067350 mL
Catalog number 20673
Price (EUR)
78,75
Each
-
Add to cart
Quantity:
50 mL
Request bulk or custom format
Price (EUR)
78,75
Each
Add to cart
Ask our AI about this Product
Thermo Scientific Pierce Dimethylformamide (DMF) is a sequencing-grade preparation with high purity and consistent absorptivity properties for use in HPLC and mass spectrometry methods.

This sequencing-grade dimethylformamide is specially purified by a proprietary method and tested to ensure lot-to-lot consistency with a low UV absorbance to provide you with the most sensitive detection across all wavelengths. DMF is packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps for ultimate protection.

Features of dimethylformamide:

• Highly purified and tested to ensure reliable performance in HPLC, mass spectrometry and sequencing applications
• Packaged in solvent-rinsed amber glass bottles and sealed with PTFE-lined fluorocarbon caps

General properties of DMF
• Alternative names: N,N-Dimethylformamide, N-Formyldimethylamine
• Molecular formula: C3H7ON
• Molecular weight: 73.09
• Density: 0.945 g/mL
• CAS number: 68-12-2

Specifications of Thermo Scientific Pierce DMF
• Particulates: Free from particulate matter
• Purity: >99.5%
• Water content: < 0.1%
• Peroxides: No peroxides detected
• Tollen's test: No aldehydes detected
• Ninhydrin positives: A570 < 0.05 above blank
• UV cutoff: < 270 nm

Pierce sequencing-grade DMF is specially purified and tested to the highest specifications to ensure the integrity of your data, maximize sensitivity in your assay and to prolong the life of your equipment.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity50 mL
For Use WithHPLC, Mass Spectrometry
FormLiquid
Item NameDimethylformamide (DMF)
Product LinePierce™
Reagent TypeSolvent (Pure)
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3239624Certificate of AnalysisJul 01, 202520673
3230798Certificate of AnalysisMay 23, 202520673
3215355Certificate of AnalysisMay 15, 202520673
AB402112Certificate of AnalysisFeb 21, 202520673
ZJ397027Certificate of AnalysisOct 31, 202420673
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link