Calcium chloride, ACS reagent, desiccant
Calcium chloride, ACS reagent, desiccant
Calcium chloride, ACS reagent, desiccant
Calcium chloride, ACS reagent, desiccant
Thermo Scientific Chemicals

Calcium chloride, ACS reagent, desiccant

CAS: 10043-52-4 | CaCl2 | 110.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 206791000
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
81.60
Save 14.95 (18%)
Each
In stock
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)
66.65
Special offer
Online exclusive
Ends: 26-Sep-2025
81.60
Save 14.95 (18%)
Each
Add to cart
Chemical Identifiers
CAS10043-52-4
IUPAC Namecalcium dichloride
Molecular FormulaCaCl2
InChI KeyUXVMQQNJUSDDNG-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Ca++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration Complexometric>=96.0 %
Appearance (Form)Powder
Titrable base=<0.006 meq/g
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0467486Certificate of AnalysisJan 30, 2025206791000, 206795000
A0466565Certificate of AnalysisOct 10, 2024206791000, 206795000
A0464155Certificate of AnalysisJul 04, 2024206791000, 206795000
A0462727Certificate of AnalysisMay 22, 2024206791000, 206795000
A0452094Certificate of AnalysisApr 15, 2024206791000, 206795000
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection