Thermo Scientific Chemicals

Lithium borohydride, 95%, Thermo Scientific Chemicals

Catalog number: 206810100
10 g, Each
Thermo Scientific Chemicals

Lithium borohydride, 95%, Thermo Scientific Chemicals

Catalog number: 206810100
10 g, Each
Quantity
Catalog number: 206810100
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16949-15-8
IUPAC Name
lithium(1+) boranuide
Molecular Formula
BH4Li
InChI Key
UUKMSDRCXNLYOO-UHFFFAOYSA-N
SMILES
[Li+].[BH4-]
Appearance (Form)
Powder
Appearance (Color)
White
Assay
>=94 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text