Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals
Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Allylpalladium chloride dimer, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 206835000
Price (USD)/ Each
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12012-95-2
IUPAC NameAllylpalladium chloride dimer
Molecular FormulaC6H10Cl2Pd2
InChI KeyPENAXHPKEVTBLF-UHFFFAOYSA-L
SMILESCl[Pd].Cl[Pd].c:c:c.c:c:c
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Appearance (Color)Yellow to yellow-green
Assay>=56.7 % (Pd)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only