Thermo Scientific Chemicals

Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206940010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Lithium tetraborate, 98+%, for analysis, Thermo Scientific Chemicals

Catalog number: 206940010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 206940010
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12007-60-2
IUPAC Name
dilithium(1+) bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
Molecular Formula
B4Li2O7
InChI Key
RDZCISUGRQUANA-UHFFFAOYSA-N
SMILES
[Li+].[Li+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2
Phosphate (PO4)
=<20 ppm
Heavy metals (as Pb)
=<50 ppm
Iron (Fe)
=<10 ppm
Sodium (Na)
=<50 ppm
Chloride (Cl)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text