Specifications
Chemical Name or MaterialL(+)-Cysteine hydrochloride monohydrate
Melting Point168°C to 170°C
CAS7048-04-6
pH0.8 to 1.2 (100 g/L, H2O, 20°C)
ColorWhite
Molecular FormulaHSCH2CH(NH2)COOH·HCl·H2O
PackagingPoly bottle, tamper-evident cap
Synonym(R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, (R)-(+)-Cysteine hydrochloride monohydrate, Cysteine Hydrochloride monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, monohydrate
Molecular Weight (g/mol)175.64
Percent Purity98.5 to 101.5%
GradeMulti-Compendial/USP
Quantity100 g
Unit Size100 g