DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
Thermo Scientific Chemicals

DL-Propranolol hydrochloride, 99%

CAS: 318-98-9 | C16H21NO2·HCl | 295.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 207320050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS318-98-9
IUPAC Namehydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Molecular FormulaC16H22ClNO2
InChI KeyZMRUPTIKESYGQW-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder
Melting point163°C to 166°C
Appearance (Color)White to light yellow
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link