DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
Thermo Scientific Chemicals

DL-Propranolol hydrochloride, 99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 207321000
Price (USD)/ Each
756.00
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
756.00
Add to cart
DL-Propranolol hydrochloride, 99%
Catalog number207321000
Price (USD)/ Each
756.00
-
Add to cart
Chemical Identifiers
CAS318-98-9
IUPAC Namehydrogen 1-(naphthalen-1-yloxy)-3-[(propan-2-yl)amino]propan-2-ol chloride
Molecular FormulaC16H22ClNO2
InChI KeyZMRUPTIKESYGQW-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(C)NCC(O)COC1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)White to light yellow
Titration with HClO4>=98.5 %
Appearance (Form)Powder
Melting point163°C to 166°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only