Salicylaldoxime, 98%, Thermo Scientific Chemicals
Salicylaldoxime, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 207401000
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
Add to cart
Salicylaldoxime, 98%, Thermo Scientific Chemicals
Catalog number207401000
Price (USD)/ Each
104.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
123.00 
Save 18.35 (15%)
-
Add to cart
Chemical Identifiers
CAS94-67-7
IUPAC Name(6Z)-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular FormulaC7H7NO2
InChI KeyGFCNBJDXQMZOOC-WAYWQWQTSA-N
SMILESON\C=C1\C=CC=CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Powder
Melting point54°C to 59°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only