Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Catalog number: 207405000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Salicylaldoxime, 98%, Thermo Scientific Chemicals

Catalog number: 207405000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207405000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
94-67-7
IUPAC Name
(6Z)-6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Molecular Formula
C7H7NO2
InChI Key
GFCNBJDXQMZOOC-WAYWQWQTSA-N
SMILES
ON\C=C1\C=CC=CC1=O
Appearance (Color)
White to light beige
Infrared spectrum
Conforms
GC
>=97.5 %
Appearance (Form)
Powder
Melting point
54°C to 59°C

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text