Thermo Scientific Chemicals

DL-Serine methyleester hydrochloride, 99%

Catalog number: 207450100
10 g, Glass Bottle, Each
Thermo Scientific Chemicals

DL-Serine methyleester hydrochloride, 99%

Catalog number: 207450100
10 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 207450100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
5619-04-5
IUPAC Name
hydrogen methyl 2-amino-3-hydroxypropanoate chloride
Molecular Formula
C4H10ClNO3
InChI Key
NDBQJIBNNUJNHA-UHFFFAOYNA-N
SMILES
[H+].[Cl-].COC(=O)C(N)CO
Appearance (Form)
Crystalline powder
Titration Argentometric
>=98.5 %
Appearance (Color)
White to off-white
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text