Thermo Scientific Chemicals

Spironolactone, 99%

Catalog number: 207460050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Spironolactone, 99%

Catalog number: 207460050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207460050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
52-01-7
IUPAC Name
(1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
Molecular Formula
C24H32O4S
InChI Key
LXMSZDCAJNLERA-ZHYRCANASA-N
SMILES
CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12
Loss on drying
=<0.5 % (105°C, 2 h)
Specific optical rotation
-41° to -45° (20°C, 589 nm) (c=1, EtOH)
Infrared spectrum
Conforms
HPLC
>=98.5 %
Appearance (Form)
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text