Thermo Scientific Chemicals

Ethyl stearate, 97%, Thermo Scientific Chemicals

Catalog number: 207480050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Ethyl stearate, 97%, Thermo Scientific Chemicals

Catalog number: 207480050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207480050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
111-61-5
IUPAC Name
ethyl octadecanoate
Molecular Formula
C20H40O2
InChI Key
MVLVMROFTAUDAG-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCC(=O)OCC
Appearance (Form)
Powder or low melting crystalline mass or liquid
Infrared spectrum
Conforms
Melting point
30°C to 39°C
Appearance (Color)
White to light yellow
GC
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text