Thiocarbohydrazide, 98%, Thermo Scientific Chemicals
Thiocarbohydrazide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thiocarbohydrazide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Plastic bottle
Catalog number 207531000
Price (USD)/ Each
475.00
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
475.00
Add to cart
Thiocarbohydrazide, 98%, Thermo Scientific Chemicals
Catalog number207531000
Price (USD)/ Each
475.00
-
Add to cart
Chemical Identifiers
CAS2231-57-4
IUPAC Name1,3-diaminothiourea
Molecular FormulaCH6N4S
InChI KeyLJTFFORYSFGNCT-UHFFFAOYSA-N
SMILESNNC(=S)NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to grey
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point168°C to 172°C (Dec)
Assay>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only