Chemical Identifiers
CAS2580-78-1
IUPAC Namedisodium 1-amino-9,10-dioxo-4-({3-[2-(sulfonatooxy)ethanesulfonyl]phenyl}amino)-9,10-dihydroanthracene-2-sulfonate
Molecular FormulaC22H16N2Na2O11S3
InChI KeyKUIXZSYWBHSYCN-UHFFFAOYSA-L
SMILES[Na+].[Na+].NC1=C2C(=O)C3=CC=CC=C3C(=O)C2=C(NC2=CC=CC(=C2)S(=O)(=O)CCOS([O-])(=O)=O)C=C1S([O-])(=O)=O
View more