Thermo Scientific Chemicals

N-(Trimethylsilyl)acetamide, 97%, Thermo Scientific Chemicals

Catalog number: 207591000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

N-(Trimethylsilyl)acetamide, 97%, Thermo Scientific Chemicals

Catalog number: 207591000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207591000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13435-12-6
IUPAC Name
N-(trimethylsilyl)acetamide
Molecular Formula
C5H13NOSi
InChI Key
LWFWUJCJKPUZLV-UHFFFAOYSA-N
SMILES
CC(=O)N[Si](C)(C)C
Melting point
40°C to 46°C
GC
>=96.0 %
Appearance (Color)
White to yellow
Appearance (Form)
Crystalline mass
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text