Thermo Scientific Chemicals

Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals

Catalog number: 207601000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals

Catalog number: 207601000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207601000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
78-50-2
IUPAC Name
1-(dioctylphosphoryl)octane
Molecular Formula
C24H51OP
InChI Key
ZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILES
CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
Infrared spectrum
Conforms
Appearance (Form)
Powder or crystals or flakes and/or chunks
Melting point
50°C to 55°C
Appearance (Color)
White to light yellow
Isomer
=<6 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text