Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals
Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 207605000
Price (USD)/ Each
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name1-(dioctylphosphoryl)octane
Molecular FormulaC24H51OP
InChI KeyZMBHCYHQLYEYDV-UHFFFAOYSA-N
SMILESCCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC
Molecular Weight (g/mol)386.65
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Powder or crystals or flakes and/or chunks
Melting point50°C to 55°C
Appearance (Color)White to light yellow
Isomer=<6 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only