Thermo Scientific Chemicals

Tungstic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 207611000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Tungstic acid, 99+%, Thermo Scientific Chemicals

Catalog number: 207611000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207611000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-03-1
IUPAC Name
dioxotungstendiol
Molecular Formula
H2O4W
InChI Key
CMPGARWFYBADJI-UHFFFAOYSA-L
SMILES
O[W](O)(=O)=O
Appearance (Form)
Powder
Assay
>=99.0 %
Appearance (Color)
Yellow to yellow-green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text