Thermo Scientific Chemicals

Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 207621000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Sodium tungstate dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 207621000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 207621000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10213-10-2
IUPAC Name
disodium dioxotungstenbis(olate) dihydrate
Molecular Formula
H4Na2O6W
InChI Key
WPZFLQRLSGVIAA-UHFFFAOYSA-N
SMILES
O.O.[Na+].[Na+].[O-][W]([O-])(=O)=O
Assay
>=99.0 %
Heavy metals (as Pb)
=<10 ppm
Free alkali (NaOH)
=<0.05 %
Arsenic (As)
=<5 ppm
Appearance (Color)
White

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text