Xanthosine dihydrate, 99%
Xanthosine dihydrate, 99%
Thermo Scientific Chemicals

Xanthosine dihydrate, 99%

Have Questions?
Quantity:
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 207630050
Price (USD)/ Each
174.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
174.00
Add to cart
Xanthosine dihydrate, 99%
Catalog number207630050
Price (USD)/ Each
174.00
-
Add to cart
Chemical Identifiers
IUPAC Name9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,3,6,9-tetrahydro-1H-purine-2,6-dione
Molecular FormulaC10H12N4O6
InChI KeyUBORTCNDUKBEOP-UUOKFMHZSA-N
SMILESOC[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=NC2=C1NC(=O)NC2=O
Molecular Weight (g/mol)284.23
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to off-white
Appearance (Form)Powder
Water9 to 14 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only