Thermo Scientific Chemicals

Zinc acetate dihydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 207645000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Zinc acetate dihydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog number: 207645000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207645000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5970-45-6
IUPAC Name
zinc(2+) diacetate dihydrate
Molecular Formula
C4H10O6Zn
InChI Key
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
SMILES
O.O.[Zn++].CC([O-])=O.CC([O-])=O
Lead (Pb)
=<50 ppm
Insoluble matter
=<0.005 %
Iron (Fe)
=<100 ppm
Titration Complexometric
>=97.5 %
Appearance (Color)
Colorless to white

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text