Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 207681000
100 g, Plastic bottle, Each
Thermo Scientific Chemicals

Copper(II) nitrate trihydrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 207681000
100 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207681000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10031-43-3
IUPAC Name
copper(2+) dinitrate
Molecular Formula
CuN2O6
InChI Key
XTVVROIMIGLXTD-UHFFFAOYSA-N
SMILES
[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Blue
Iron (Fe)
=<50 ppm
Appearance (Form)
Powder or crystals and/or chunks
Chloride (Cl)
=<20 ppm
Lead (Pb)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text