Sodium peroxide, 96%, Thermo Scientific Chemicals
Sodium peroxide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium peroxide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
100 g
500 g
Packaging:
Plastic bottle
Catalog number 207700025
Price (USD)/ Each
1,401.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1649.00 
Save 247.35 (15%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
1,401.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1649.00 
Save 247.35 (15%)
Add to cart
Sodium peroxide, 96%, Thermo Scientific Chemicals
Catalog number207700025
Price (USD)/ Each
1,401.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
1649.00 
Save 247.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium dioxidanediide
Molecular FormulaNa2O2
InChI KeyPFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES[Na+].[Na+].[O-][O-]
Molecular Weight (g/mol)77.98
View more
SpecificationsSpecification SheetSpecification Sheet
Appearancematerial from the manufacturing process
Sulfate (SO4)=<50 ppm
Appearance (Color)Yellow
AppearanceMay contain a small quantity of discolored
Titration with KMnO4>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only