Thermo Scientific Chemicals

Sodium peroxide, 96%, Thermo Scientific Chemicals

Catalog number: 207700250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Sodium peroxide, 96%, Thermo Scientific Chemicals

Catalog number: 207700250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207700250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1313-60-6
IUPAC Name
disodium dioxidanediide
Molecular Formula
Na2O2
InChI Key
PFUVRDFDKPNGAV-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][O-]
Appearance (Form)
Granules
Appearance
material from the manufacturing process
Sulfate (SO4)
=<50 ppm
Appearance (Color)
Yellow
Appearance
May contain a small quantity of discolored

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text