Thermo Scientific Chemicals

Potassium sulfate, 99+%, for analysis, anhydrous powder, Thermo Scientific Chemicals

Catalog number: 207775000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium sulfate, 99+%, for analysis, anhydrous powder, Thermo Scientific Chemicals

Catalog number: 207775000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207775000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7778-80-5
IUPAC Name
dipotassium sulfate
Molecular Formula
K2O4S
InChI Key
OTYBMLCTZGSZBG-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S([O-])(=O)=O
Appearance (Color)
White
Heavy metals (as Pb)
=<5 ppm
Total nitrogen (as N)
=<0.0005 %
Iron (Fe)
=<5 ppm
Calcium (Ca)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text