Thermo Scientific Chemicals

Calcium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 207780100
10 kg, Plastic drum, Each
Thermo Scientific Chemicals

Calcium chloride dihydrate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 207780100
10 kg, Plastic drum, Each
Quantity
Packaging
Catalog number: 207780100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10035-04-8
IUPAC Name
calcium dihydrate dichloride
Molecular Formula
CaCl2H4O2
InChI Key
LLSDKQJKOVVTOJ-UHFFFAOYSA-L
SMILES
O.O.[Cl-].[Cl-].[Ca++]
Titration Complexometric
>=99.0 %
Iron (Fe)
=<5 ppm
Appearance (Form)
Crystalline powder or crystals
Heavy metals (as Pb)
=<5 ppm
Sulfate (SO4)
=<100 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text