Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Sodium phosphate, monobasic monohydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium phosphate, monobasic monohydrate, 99+%, for analysis

CAS: 10049-21-5 | H5NaO5P | 139.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207805000
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
132.65
Online Exclusive
147.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10049-21-5
IUPAC Namesodium phosphoric acid hydrate
Molecular FormulaH5NaO5P
InChI KeyBBMHARZCALWXSL-UHFFFAOYSA-N
SMILESO.[Na+].OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Sulfate (SO4)=<30 ppm
Insoluble matter=<0.01 % (Ca and NH4OH ppt)
Chloride (Cl)=<5 ppm
Appearance (Color)White
Titration with NaOH>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only

Share catalog number, name or link