2079-HR Full specifications
Case of 3072 ART Barrier, Filtered, Sterile, Hinged Rack 1000 μL 11.85cm Sterile & PCR Ready 510.30 Online price 729.00
- Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 1000 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 4.7 in. Length (Metric) 11.85cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 3072
2140-HR Full specifications
Case of 4800 ART 10 REACH, Filtered, Sterile, Hinged Rack 10 μL 4.33cm Sterile & PCR Ready 620.90 Online price 887.00
- Description ART 10 REACH, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 REACH Length (English) 1.71 in. Length (Metric) 4.33cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Extended Length For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2140-HRPK Full specifications
Pack of 960 ART 10 REACH, Filtered, Sterile, Hinged Rack 10 μL 4.33cm Sterile & PCR Ready - Description ART 10 REACH, Filtered, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 REACH Length (English) 1.71 in. Length (Metric) 4.33cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Extended Length For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2140-05-HR Full specifications
Case of 4800 ART 10 REACH, Filtered, LR, Sterile, Hinged Rack 10 μL 4.33cm Sterile & PCR Ready 694.40 Online price 992.00
- Description ART 10 REACH, Filtered, LR, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 REACH Length (English) 1.71 in. Length (Metric) 4.33cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 4800
2140-05-HRPK also known as 2140-05-HR
Full specifications
Pack of 960 ART 10 REACH, Filtered, LR, Sterile, Hinged Rack 10 μL 4.33cm Sterile & PCR Ready - Description ART 10 REACH, Filtered, LR, Sterile, Hinged Rack Volume (Metric) 10 μL Filtered Filtered Sterility Sterile Model ART 10 REACH Length (English) 1.71 in. Length (Metric) 4.33cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 960
2160P-HR Full specifications
Case of 3840 ART Barrier, Filtered, Sterile, Hinged Rack 200 μL 9.09cm Sterile & PCR Ready 666.40 Online price 952.00
- Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 3.6 in. Length (Metric) 9.09cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 3840
2160P-HRPK also known as 2160P-HR
Full specifications
Pack of 768 ART Barrier, Filtered, Sterile, Hinged Rack 200 μL 9.09cm Sterile & PCR Ready - Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 3.6 in. Length (Metric) 9.09cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 768
2160P-05-HR Full specifications
Case of 3840 ART Barrier, Filtered, Sterile, Hinged Rack 200 μL 9.09cm Sterile & PCR Ready - Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 3.6 in. Length (Metric) 9.09cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Case of 3840
2160P-05-HRPK also known as 2160P-05-HR
Full specifications
Pack of 768 ART Barrier, Filtered, Sterile, Hinged Rack 200 μL 9.09cm Sterile & PCR Ready - Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 200 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 3.6 in. Length (Metric) 9.09cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 768
2079-HRPK Full specifications
Pack of 768 ART Barrier, Filtered, Sterile, Hinged Rack 1000 μL 11.85cm Sterile & PCR Ready - Description ART Barrier, Filtered, Sterile, Hinged Rack Volume (Metric) 1000 μL Filtered Filtered Sterility Sterile Color Natural Model ART Barrier Length (English) 4.7 in. Length (Metric) 11.85cm Clean Claims DNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free Format Hinged Rack Tip Style Universal Fit For Use With (Equipment) Universal Fit Product Line ART Fit for Purpose Sterile & PCR Ready Unit Size Pack of 768