Thermo Scientific Chemicals

Potassium pyrosulfate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 207915000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium pyrosulfate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 207915000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 207915000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-62-7
IUPAC Name
dipotassium (sulfonatooxy)sulfonate
Molecular Formula
K2O7S2
InChI Key
KAQHZJVQFBJKCK-UHFFFAOYSA-L
SMILES
[K+].[K+].[O-]S(=O)(=O)OS([O-])(=O)=O
Appearance (Form)
Crystalline powder
Appearance (Color)
White
Phosphate (PO4)
=<10 ppm
Heavy metals (as Pb)
=<10 ppm
Iron (Fe)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text