Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals
Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 207991000
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
-
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
Add to cart
Vanadium(V) oxide, 98+%, Thermo Scientific Chemicals
Catalog number207991000
Price (USD)/ Each
28.65
Online exclusive
31.70 
Save 3.05 (10%)
-
Add to cart
Chemical Identifiers
CAS1314-62-1
IUPAC Name[(dioxovanadio)oxy]dioxovanadium
Molecular FormulaO5V2
InChI KeyGNTDGMZSJNCJKK-UHFFFAOYSA-N
SMILESO=[V](=O)O[V](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Sodium oxide (Na2O)=<300 ppm
Iron (Fe)=<1000 ppm
Chromium (Cr)=<200 ppm
Assay>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only