Thermo Scientific Chemicals

Barium iodide hydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 208000500
50 g, Glass bottle, Each
Thermo Scientific Chemicals

Barium iodide hydrate, 98%, for analysis, Thermo Scientific Chemicals

Catalog number: 208000500
50 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208000500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
85017-89-6
IUPAC Name
barium(2+) diiodide
Molecular Formula
BaI2
InChI Key
SGUXGJPBTNFBAD-UHFFFAOYSA-L
SMILES
[I-].[I-].[Ba++]
Appearance (Color)
White to off-white
Titration Argentometric
>=97.5 % (on dry substance)
Iron (Fe)
=<5 ppm
Chromium (Cr)
=<10 ppm
Manganese (Mn)
=<10 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text