1x1 image pixel for data collection
Zinc iodide, 98+%, pure
Zinc iodide, 98+%, pure
Zinc iodide, 98+%, pure
Zinc iodide, 98+%, pure
Thermo Scientific Chemicals

Zinc iodide, 98+%, pure

CAS: 10139-47-6 | I2Zn | 319.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 208062500
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
145.65
Online Exclusive
162.00
Save 16.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS10139-47-6
IUPAC Namezinc(2+) diiodide
Molecular FormulaI2Zn
InChI KeyUAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES[Zn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to light beige
Titration Argentometric>=98.0 %
Appearance (Form)Hygroscopic crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link