Thermo Scientific Chemicals

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, Thermo Scientific Chemicals

Catalog number: 208090025
2.5 kg, Glass Bottle, Each
Thermo Scientific Chemicals

Ascarite(II), CO2 absorbent, 20-30 mesh, capacity: 40-50%, Thermo Scientific Chemicals

Catalog number: 208090025
2.5 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 208090025
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81133-20-2, 1310-73-2, 7631-86-9
Particle size
20 to 30 Mesh
Appearance (Form)
Granules
Appearance (Color)
Beige to light brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Gas Adsorbent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text