L-Carnosine, 98%
Thermo Scientific Chemicals

L-Carnosine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 208171000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS305-84-0
IUPAC Name(2S)-2-(3-aminopropanamido)-3-(1H-imidazol-5-yl)propanoic acid
Molecular FormulaC9H14N4O3
InChI KeyCQOVPNPJLQNMDC-ZETCQYMHSA-N
SMILESNCCC(=O)N[C@@H](CC1=CN=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Specific optical rotation+20.0° to +22.0° (20°C, 589 nm) (c=2, water)
Water=<1 %
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only