Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Phosphotungstic acid hydrate, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 208310250
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
Add to cart
Phosphotungstic acid hydrate, Thermo Scientific Chemicals
Catalog number208310250
Price (USD)/ Each
64.65
Online exclusive
71.20 
Save 6.55 (9%)
-
Add to cart
Chemical Identifiers
CAS12501-23-4
IUPAC Name12-Tungstophosphate hydrate
Molecular Weight (g/mol)2880.20
MDL NumberMFCD00011342
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to grey
Assay>=82 % (WO3)
Chloride (Cl)=<20 ppm
Iron (Fe)=<30 ppm
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only