Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Thermo Scientific Chemicals

Phosphotungstic acid hydrate

CAS: 12501-23-4 | H5O41PW12 | 2880.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 208315000
Price (USD)
639.65
Special offer
Online exclusive
752.00
Save 112.35 (15%)
Each
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
639.65
Special offer
Online exclusive
752.00
Save 112.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS12501-23-4
IUPAC Name12-Tungstophosphate hydrate
Molecular Weight (g/mol)2880.20
MDL NumberMFCD00011342
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)=<50 ppm
Sodium (Na)=<100 ppm
Iron (Fe)=<30 ppm
Appearance (Color)White to yellow to beige to grey
Assay>=82 % (WO3)
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection