2-Propylpentanoic acid, 99%, Thermo Scientific Chemicals
2-Propylpentanoic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Propylpentanoic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 208330250
Price (USD)/ Each
56.65
Online exclusive
67.00 
Save 10.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
56.65
Online exclusive
67.00 
Save 10.35 (15%)
Add to cart
2-Propylpentanoic acid, 99%, Thermo Scientific Chemicals
Catalog number208330250
Price (USD)/ Each
56.65
Online exclusive
67.00 
Save 10.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Name2-propylpentanoic acid
Molecular FormulaC8H16O2
InChI KeyNIJJYAXOARWZEE-UHFFFAOYSA-N
SMILESCCCC(CCC)C(O)=O
Molecular Weight (g/mol)144.21
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4240 to 1.4260 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only