Thermo Scientific Chemicals

Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 208375000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Zirconyl chloride octahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 208375000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208375000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13520-92-8
Molecular Formula
Cl2H18O9Zr
InChI Key
FUBACIUATZGHAC-UHFFFAOYSA-N
SMILES
O.O.O.O.O.O.O.O.Cl.Cl.O=[Zr]
Molecular Weight (g/mol)
324.26
Appearance (Color)
White to light yellow
Appearance (Form)
Crystalline powder
Water
42 to 50 %
Titration Argentometric
97.5 to 102.5 %
Solubility
(10% in water) Clear to slightly hazy solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text