Thermo Scientific Chemicals

Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 208461000
100 mL, AcroSeal™ Glass Bottle, Each
Thermo Scientific Chemicals

Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Catalog number: 208461000
100 mL, AcroSeal™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 208461000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
32916-51-1
IUPAC Name
chloro(cyclopentyl)magnesium
Molecular Formula
C5H9ClMg
InChI Key
GROGEABZCKQTJW-UHFFFAOYSA-M
SMILES
Cl[Mg]C1CCCC1
Titration Grignard
1.9 to 2.2 M
Appearance (Color)
Clear to hazy yellow to grey to brown
Appearance
May develop some turbidity or precipitate
Appearance (Form)
Solution

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text