Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals
Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 208745000
Price (USD)/ Each
49.65
Online exclusive
58.20 
Save 8.55 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)/ Each
49.65
Online exclusive
58.20 
Save 8.55 (15%)
Add to cart
Succinic acid, disodium salt, 99%, anhydrous, Thermo Scientific Chemicals
Catalog number208745000
Price (USD)/ Each
49.65
Online exclusive
58.20 
Save 8.55 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namedisodium butanedioate
Molecular FormulaC4H4Na2O4
InChI KeyZDQYSKICYIVCPN-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)CCC([O-])=O
Molecular Weight (g/mol)162.05
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HClO4>=98.5 %
Appearance (Form)Crystalline powder
Loss on drying=<0.3 % (120°C)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only