Thermo Scientific Chemicals

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 208750050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Tris(2,2'-bipyridyl)ruthenium(II) chloride, hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: 208750050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208750050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
50525-27-4
IUPAC Name
λ²-ruthenium(2+) tris(2,2'-bipyridine) hexahydrate dichloride
Molecular Formula
C30H36Cl2N6O6Ru
InChI Key
WHELTKFSBJNBMQ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.[Cl-].[Cl-].[Ru++].C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1.C1=CN=C(C=C1)C1=NC=CC=C1
Appearance (Color)
Orange to red
Elemental Analysis
>=97.5 %
Appearance (Form)
Crystalline powder or crystals

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text