Thermo Scientific Chemicals

Methyl orotate, 98%, ACROS Organics™

Catalog number: 208781000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Methyl orotate, 98%, ACROS Organics™

Catalog number: 208781000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208781000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6153-44-2
IUPAC Name
methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Molecular Formula
C6H6N2O4
InChI Key
UUTDWTOZAWFKFW-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(=O)NC(=O)N1
Appearance (Form)
Powder
Appearance (Color)
White to yellow to light brown
Titration with NaOH
94.0 to 106.0 %
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text