Thermo Scientific Chemicals

Bismuth(III) chloride, 97+%, anhydrous, Thermo Scientific Chemicals

Catalog number: 208831000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Bismuth(III) chloride, 97+%, anhydrous, Thermo Scientific Chemicals

Catalog number: 208831000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208831000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7787-60-2
IUPAC Name
bismuth(3+) trichloride
Molecular Formula
BiCl3
InChI Key
JHXKRIRFYBPWGE-UHFFFAOYSA-K
SMILES
[Cl-].[Cl-].[Cl-].[Bi+3]
Appearance (Form)
Crystalline powder and/or chunks
Titration Argentometric
>=98.0 %
Appearance (Color)
White to beige to grey
Water
=<3 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text