Thermo Scientific Chemicals

Iodine monobromide, 98%, Thermo Scientific Chemicals

Catalog number: 208842500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodine monobromide, 98%, Thermo Scientific Chemicals

Catalog number: 208842500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 208842500
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7789-33-5
IUPAC Name
iodobromane
Molecular Formula
BrI
InChI Key
CBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILES
BrI
Appearance (Form)
Crystals or crystalline mass and/or chunks
Appearance (Color)
Dark grey to brown-black
Sulfated ash
=<0.05 %
Titration Iodometric
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text