Thermo Scientific Chemicals

Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 208860010
1 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Potassium chlorate, 99%, extra pure, Thermo Scientific Chemicals

Catalog number: 208860010
1 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 208860010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
3811-04-9
IUPAC Name
potassium chlorate
Molecular Formula
ClKO3
InChI Key
VKJKEPKFPUWCAS-UHFFFAOYSA-M
SMILES
[K+].[O-][Cl](=O)=O
Chloride (Cl)
=<2000 ppm
Chromium (Cr)
=<200 ppm
Appearance (Form)
Crystalline powder
Assay
>=98.5 % (KClO3)
Insoluble matter
=<0.2 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text