Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals
Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Glass bottle
Catalog number 208875000
Price (USD)/ Each
82.65
Online exclusive
97.10 
Save 14.45 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
82.65
Online exclusive
97.10 
Save 14.45 (15%)
Add to cart
Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals
Catalog number208875000
Price (USD)/ Each
82.65
Online exclusive
97.10 
Save 14.45 (15%)
-
Add to cart
Chemical Identifiers
IUPAC Namesodium bromate
Molecular FormulaBrNaO3
InChI KeyXUXNAKZDHHEHPC-UHFFFAOYSA-M
SMILES[Na+].[O-][Br](=O)=O
Molecular Weight (g/mol)150.89
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<50 ppm
Bromide (Br)=<500 ppm
Titration Iodometric>=99.0 %
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only