Thallium(I) chloride, 99%, Thermo Scientific Chemicals
Thallium(I) chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thallium(I) chloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 208880250
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
Add to cart
Thallium(I) chloride, 99%, Thermo Scientific Chemicals
Catalog number208880250
Price (USD)/ Each
90.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
107.00 
Save 16.35 (15%)
-
Add to cart
Chemical Identifiers
CAS7791-12-0
IUPAC Nameλ¹-thallanylium chloride
Molecular FormulaClTl
InChI KeyGBECUEIQVRDUKB-UHFFFAOYSA-M
SMILES[Cl-].[Tl+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration Argentometric98.5 to 101.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only